Assay Details
Target Gene Details
Entrez Gene ID: | 79258 |
Gene Name: | membrane metallo-endopeptidase-like 1 |
Gene Aliases: |
MMEL2, NEP2, NEPII, NL1, NL2, SEP |
Location: |
Chr.1:2590639-2633042 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMEL1 | NM_033467.3 | 20 | 2168 | NP_258428.2 |
| XM_011542122.2 | 13 | 1271 | XP_011540424.1 | |
| XM_017002310.1 | 21 | 2434 | XP_016857799.1 | |
| XM_017002311.1 | 20 | 2114 | XP_016857800.1 | |
| XM_017002312.1 | 19 | 2086 | XP_016857801.1 | |
| XM_017002313.1 | 20 | 2168 | XP_016857802.1 | |
| XM_017002314.1 | 19 | 2039 | XP_016857803.1 | |
| XM_017002315.1 | 20 | 2044 | XP_016857804.1 | |
| AF336981.1 | 19 | 1991 | AAL08942.1 | |
| AK093058.1 | 20 | 2116 | ||
| BC101027.2 | 19 | 1964 | ||
| BC101028.1 | 15 | 1493 | ||
| BC101029.2 | 19 | 1993 | AAI01030.1 | |
| BC101030.1 | 19 | 1964 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829648 | Chr.1:2567689 - 2640901 on Build GRCh38 | Loss |
|
| esv3582248 | Chr.1:2569899 - 2636548 on Build GRCh38 | Gain |
|
| nsv545077 | Chr.1:2428542 - 2622185 on Build GRCh38 | Gain |
|
| nsv545082 | Chr.1:2536231 - 2597243 on Build GRCh38 | Loss |
|
| nsv470684 | Chr.1:2394570 - 2930923 on Build GRCh38 | Loss |
|
| nsv951592 | Chr.1:2564862 - 2613961 on Build GRCh38 | Deletion |
|
| nsv545083 | Chr.1:2575976 - 2653099 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map