Assay Details
Target Gene Details
Entrez Gene ID: | 7224 |
Gene Name: | transient receptor potential cation channel subfamily C member 5 |
Gene Aliases: |
PPP1R159, TRP5 |
Location: |
Chr.X:111774314-112082943 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC5 | NM_012471.2 | 11 | 3656 | NP_036603.1 |
| XM_017029774.1 | 12 | 3946 | XP_016885263.1 | |
| AF054568.1 | 11 | 3655 | AAF00002.1 | |
| BC137271.1 | 11 | 3496 | ||
| BC137274.1 | 11 | 3496 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7039 | Chr.X:111761020 - 111806086 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map