Assay Details
Target Gene Details
Entrez Gene ID: | 25917 |
Gene Name: | THUMP domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.3:9362842-9386791 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
More Information
Set Membership: |
|

Genomic Map