Assay Details
Target Gene Details
Entrez Gene ID: | 116840 |
Gene Name: | centrobin, centriole duplication and spindle assembly protein |
Gene Aliases: |
LIP8, PP1221 |
Location: |
Chr.17:7932054-7949919 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574333 | Chr.17:7880977 - 7952801 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map