Assay Details
Target Gene Details
Entrez Gene ID: | 55862 |
Gene Name: | ethylmalonyl-CoA decarboxylase 1 |
Gene Aliases: |
HEL-S-76, MMCD, dJ351K20.2 |
Location: |
Chr.6:127288712-127343609 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECHDC1 | NM_001002030.1 | 6 | 1648 | NP_001002030.1 |
| NM_001105544.1 | 5 | 1426 | NP_001099014.1 | |
| NM_001105545.1 | 4 | 1373 | NP_001099015.1 | |
| NM_001139510.1 | 6 | 1426 | NP_001132982.1 | |
| NM_018479.3 | 5 | 1595 | NP_060949.2 | |
| XM_005267047.2 | 7 | 1573 | XP_005267104.1 | |
| XM_005267048.1 | 6 | 1747 | XP_005267105.1 | |
| XM_005267049.2 | 5 | 1862 | XP_005267106.1 | |
| XM_005267050.2 | 6 | 1488 | XP_005267107.1 | |
| XM_011535942.2 | 7 | 1544 | XP_011534244.1 | |
| XM_017011056.1 | 7 | 1564 | XP_016866545.1 | |
| AK025760.1 | 6 | 1398 | ||
| AK025796.1 | 6 | 1426 | ||
| AK303812.1 | 5 | 1194 | ||
| AL834469.1 | 6 | 1587 | CAD39128.1 | |
| BU684175.1 | ||||
| EU668355.1 | 4 | 1373 |
Target Gene Details
Entrez Gene ID: | 81847 |
Gene Name: | ring finger protein 146 |
Gene Aliases: |
- |
Location: |
Chr.6:127266340-127289893 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF146 | XM_006715571.3 | 5 | 3923 | XP_006715634.1 |
| XM_011536161.2 | 6 | 3810 | XP_011534463.1 | |
| XM_011536162.2 | 6 | 3862 | XP_011534464.1 | |
| XM_011536163.2 | 6 | 3842 | XP_011534465.1 | |
| XM_011536164.2 | 5 | 3701 | XP_011534466.1 | |
| XM_017011336.1 | 3 | 3603 | XP_016866825.1 | |
| XM_017011337.1 | 5 | 6844 | XP_016866826.1 | |
| XM_017011338.1 | 7 | 3951 | XP_016866827.1 | |
| XM_017011339.1 | 6 | 3980 | XP_016866828.1 | |
| XM_017011340.1 | 6 | 4032 | XP_016866829.1 | |
| XM_017011341.1 | 4 | 3731 | XP_016866830.1 | |
| XM_017011342.1 | 5 | 3872 | XP_016866831.1 | |
| XM_017011343.1 | 6 | 3981 | XP_016866832.1 |
More Information
Set Membership: |
|

Genomic Map