Assay Details
Target Gene Details
Entrez Gene ID: | 137362 |
Gene Name: | glutamic-oxaloacetic transaminase 1-like 1 |
Gene Aliases: |
- |
Location: |
Chr.8:37933548-37940502 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOT1L1 | NM_152413.2 | 9 | 1280 | NP_689626.2 |
| XM_005273399.3 | 9 | 1649 | XP_005273456.1 | |
| XM_006716285.3 | 8 | 1560 | XP_006716348.1 | |
| XM_011544391.2 | 6 | 736 | XP_011542693.1 | |
| BC029504.1 | 9 | 1281 | AAH29504.1 |
More Information
Set Membership: |
|

Genomic Map