Assay Details
Target Gene Details
Entrez Gene ID: | 81618 |
Gene Name: | integral membrane protein 2C |
Gene Aliases: |
BRI3, BRICD2C, E25, E25C, ITM3 |
Location: |
Chr.2:230864598-230879254 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITM2C | NM_001012514.2 | 5 | 1268 | NP_001012532.1 |
| NM_001012516.2 | 5 | 1243 | NP_001012534.1 | |
| NM_001287240.1 | 6 | 1345 | NP_001274169.1 | |
| NM_001287241.1 | 7 | 1541 | NP_001274170.1 | |
| NM_030926.5 | 6 | 1409 | NP_112188.1 | |
| AF271781.1 | 6 | 1394 | AAG44792.1 | |
| AF272043.1 | 6 | 1391 | AAF89492.1 | |
| AK056321.1 | 5 | 1215 | ||
| AK075361.1 | 6 | 1374 | BAC11570.1 | |
| AK090975.1 | 6 | 1320 | BAC03562.1 | |
| AK126739.1 | 1 | 2489 | ||
| AK222697.1 | 6 | 1356 | BAD96417.1 | |
| AK303199.1 | ||||
| AK307702.1 | 5 | 1177 | ||
| AL136603.1 | 6 | 1350 | CAB66538.1 | |
| AL713651.1 | 6 | 1333 | CAD28460.1 | |
| BC002424.1 | 6 | 1336 | AAH02424.1 | |
| BC025742.1 | 5 | 1213 | AAH25742.1 | |
| BC050668.1 | 5 | 1203 | AAH50668.1 | |
| BC098563.1 | 6 | 1322 | AAH98563.1 | |
| CR749389.1 | 1 | 2446 | ||
| D87062.1 | 1 | 356 | BAB46918.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470522 | Chr.2:230867985 - 230943576 on Build GRCh38 | Loss |
|
| nsv834565 | Chr.2:230697957 - 230920661 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map