Assay Details
Target Gene Details
Entrez Gene ID: | 64919 |
Gene Name: | B-cell CLL/lymphoma 11B |
Gene Aliases: |
ATL1, ATL1-alpha, ATL1-beta, ATL1-delta, ATL1-gamma, CTIP-2, CTIP2, RIT1, ZNF856B, hRIT1-alpha |
Location: |
Chr.14:99169287-99271713 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL11B | NM_001282237.1 | 4 | 7352 | NP_001269166.1 |
| NM_001282238.1 | 3 | 7139 | NP_001269167.1 | |
| NM_022898.2 | 3 | 7142 | NP_075049.1 | |
| NM_138576.3 | 4 | 7355 | NP_612808.1 | |
| AB041218.1 | 4 | 7355 | ||
| AB057668.1 | 4 | 7352 | ||
| AB057669.1 | 3 | 7142 | ||
| AB057670.1 | 3 | 7139 | ||
| AJ404614.1 | 3 | 6914 | CAC17726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv565706 | Chr.14:99156693 - 99190855 on Build GRCh38 | Loss |
|
| nsv827039 | Chr.14:99168373 - 99182672 on Build GRCh38 | Gain |
|
| esv3635480 | Chr.14:99145645 - 99269571 on Build GRCh38 | Gain |
|
| nsv565707 | Chr.14:99166653 - 99175671 on Build GRCh38 | Loss |
|
| nsv827040 | Chr.14:99169254 - 99194041 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map