Assay Details
Target Gene Details
Entrez Gene ID: | 1459 |
Gene Name: | casein kinase 2 alpha 2 |
Gene Aliases: |
CK2A2, CK2alpha', CSNK2A1 |
Location: |
Chr.16:58157907-58198116 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSNK2A2 | NM_001896.2 | NP_001887.1 | ||
| XM_005255800.3 | XP_005255857.1 | |||
| XM_005255801.3 | XP_005255858.1 | |||
| XM_017022945.1 | XP_016878434.1 | |||
| AK125922.1 | ||||
| BC008812.2 | AAH08812.1 | |||
| M55268.1 | AAA51548.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638719 | Chr.16:58084081 - 58175175 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map