Assay Details
Target Gene Details
Entrez Gene ID: | 91782 |
Gene Name: | charged multivesicular body protein 7 |
Gene Aliases: |
- |
Location: |
Chr.8:23243637-23262000 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHMP7 | NM_152272.4 | 2 | 496 | NP_689485.1 |
| XM_017013962.1 | 2 | 422 | XP_016869451.1 | |
| XM_017013964.1 | 2 | 422 | XP_016869453.1 | |
| AK097202.1 | 2 | 496 | ||
| BC042050.1 | 2 | 477 | AAH42050.1 | |
| DB140277.1 | 2 | 423 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1027718 | Chr.8:22805950 - 23258334 on Build GRCh38 | Gain |
|
| esv2759604 | Chr.8:20675533 - 23254096 on Build GRCh38 | Loss |
|
| nsv1017570 | Chr.8:22969562 - 23544783 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map