Assay Details
Target Gene Details
Entrez Gene ID: | 84668 |
Gene Name: | family with sequence similarity 126 member A |
Gene Aliases: |
DRCTNNB1A, HCC, HLD5, HYCC1 |
Location: |
Chr.7:22895848-23014195 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM126A | NM_032581.3 | 11 | 3260 | NP_115970.2 |
| XM_005249894.3 | 12 | 3600 | XP_005249951.1 | |
| XM_005249895.2 | 12 | 3519 | XP_005249952.1 | |
| XM_011515589.2 | 11 | 3111 | XP_011513891.1 | |
| XM_011515590.1 | XP_011513892.1 | |||
| AK297399.1 | ||||
| AL833296.1 | 11 | 3424 | ||
| BC018710.1 | 11 | 3238 | AAH18710.1 | |
| BX640757.1 | 1 | 3557 | CAE45864.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526131 | Chr.7:22802700 - 23199514 on Build GRCh38 | Loss |
|
| nsv1027363 | Chr.7:22817000 - 23271413 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map