Assay Details
Target Gene Details
Entrez Gene ID: | 10036 |
Gene Name: | chromatin assembly factor 1 subunit A |
Gene Aliases: |
CAF-1, CAF1, CAF1B, CAF1P150, P150 |
Location: |
Chr.19:4402596-4448322 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHAF1A |
Target Gene Details
Entrez Gene ID: | 80700 |
Gene Name: | UBX domain protein 6 |
Gene Aliases: |
UBXD1, UBXDC2 |
Location: |
Chr.19:4445006-4458759 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBXN6 | NM_001171091.1 | NP_001164562.1 | ||
| NM_025241.2 | NP_079517.1 | |||
| XM_017027325.1 | XP_016882814.1 | |||
| AF272893.1 | AAK13257.1 | |||
| AF272894.1 | AAK13258.1 | |||
| AK090473.1 | 1 | 1264 | BAC03454.1 | |
| BC007414.2 | AAH07414.2 | |||
| BC008288.1 | AAH08288.1 | |||
| BM764980.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833720 | Chr.19:4367721 - 4540556 on Build GRCh38 | Loss |
|
| nsv833718 | Chr.19:4221577 - 4480248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map