Assay Details
Target Gene Details
Entrez Gene ID: | 10220 |
Gene Name: | growth differentiation factor 11 |
Gene Aliases: |
BMP-11, BMP11 |
Location: |
Chr.12:55743278-55757194 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GDF11 | NM_005811.3 | NP_005802.1 | ||
| XM_006719194.3 | XP_006719257.1 | |||
| AF028333.1 | AAF21630.1 | |||
| AF100907.1 | AAC72852.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832423 | Chr.12:55687038 - 55859914 on Build GRCh38 | Gain |
|
| nsv1070113 | Chr.12:55743115 - 55746216 on Build GRCh38 | Deletion |
|
| nsv952185 | Chr.12:55743117 - 55746416 on Build GRCh38 | Deletion |
|
| nsv469410 | Chr.12:55704254 - 55777109 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map