Assay Details
Target Gene Details
Entrez Gene ID: | 55969 |
Gene Name: | chromosome 20 open reading frame 24 |
Gene Aliases: |
PNAS-11, RIP5 |
Location: |
Chr.20:36605734-36612557 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C20orf24 | NM_001199534.1 | NP_001186463.1 | ||
| NM_018840.4 | NP_061328.1 | |||
| NM_199483.2 | NP_955777.1 | |||
| NR_026562.3 | ||||
| AF112213.1 | AAF17201.1 | |||
| AF274936.1 | AAK07515.1 | |||
| AF274948.1 | AAK07525.1 | |||
| BC001871.1 | AAH01871.1 | |||
| BG395753.1 | ||||
| BI223439.1 | ||||
| BQ086248.1 | ||||
| BU189639.1 | ||||
| BU848918.1 | ||||
| S83364.1 | AAB50849.1 |
Target Gene Details
Entrez Gene ID: | 100527943 |
Gene Name: | TGIF2-C20orf24 readthrough |
Gene Aliases: |
- |
Location: |
Chr.20:36574554-36612557 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGIF2-C20orf24 | NM_001199535.1 | NP_001186464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss |
|
| nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map