Assay Details
Target Gene Details
Entrez Gene ID: | 9244 |
Gene Name: | cytokine receptor like factor 1 |
Gene Aliases: |
CISS, CISS1, CLF, CLF-1, NR6, zcytor5 |
Location: |
Chr.19:18593225-18606850 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRLF1 | NM_004750.4 | 6 | 1168 | NP_004741.1 |
| XM_011528422.2 | 6 | 1250 | XP_011526724.1 | |
| XM_011528423.2 | 6 | 974 | XP_011526725.1 | |
| XM_011528424.2 | 6 | 1242 | XP_011526726.1 | |
| AF059293.1 | 6 | 1092 | AAC28335.1 | |
| AF073515.1 | 6 | 1177 | AAD39681.1 | |
| AF178684.1 | 6 | 1025 | AAD54385.1 | |
| AY358291.1 | 6 | 1104 | AAQ88658.1 | |
| BC044634.1 | 6 | 1101 | AAH44634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833776 | Chr.19:18527218 - 18671454 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map