Assay Details
Target Gene Details
Entrez Gene ID: | 8909 |
Gene Name: | endonuclease, poly(U) specific |
Gene Aliases: |
P11, PP11, PRSS26 |
Location: |
Chr.12:47709734-47725572 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENDOU | NM_001172439.1 | 10 | 2406 | NP_001165910.1 |
| NM_001172440.1 | 8 | 2217 | NP_001165911.1 | |
| NM_006025.3 | 9 | 2283 | NP_006016.1 | |
| AI039727.1 | 1 | 69 | ||
| AK075446.1 | 10 | 2370 | ||
| BC069715.1 | AAH69715.1 | |||
| M32402.1 | 9 | 2278 | AAA36464.1 | |
| M36109.1 | 9 | 2278 | AAA36465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751101 | Chr.12:47547075 - 47733102 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map