Assay Details
Target Gene Details
Entrez Gene ID: | 64061 |
Gene Name: | TSPY like 2 |
Gene Aliases: |
CDA1, CINAP, CTCL, DENTT, HRIHFB2216, NP79, SE204, TSPX |
Location: |
Chr.X:53082327-53088546 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPYL2 | NM_022117.3 | 1 | 418 | NP_071400.1 |
| XM_006724592.3 | 1 | 451 | XP_006724655.1 | |
| XM_017029726.1 | 1 | 451 | XP_016885215.1 | |
| XM_017029727.1 | 1 | 451 | XP_016885216.1 | |
| AF254794.1 | 1 | 157 | AAG53596.1 | |
| AF273046.1 | 1 | 408 | AAG34906.1 | |
| AK131220.1 | 1 | 411 | ||
| AK292462.1 | 1 | 411 | ||
| AL713652.1 | 1 | 397 | CAD28461.1 | |
| AY040871.1 | 1 | 407 | AAK72407.1 | |
| BC024270.1 | 1 | 398 | AAH24270.1 | |
| DA076199.1 | 1 | 418 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv508773 | Chr.X:53034408 - 53141410 on Build GRCh38 | Insertion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map