Assay Details
Target Gene Details
Entrez Gene ID: | 9843 |
Gene Name: | hephaestin |
Gene Aliases: |
CPL |
Location: |
Chr.X:66162526-66267389 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEPH | NM_001130860.3 | NP_001124332.1 | ||
| NM_001282141.1 | NP_001269070.1 | |||
| NM_014799.3 | NP_055614.1 | |||
| NM_138737.4 | NP_620074.3 | |||
| XM_006724722.1 | XP_006724785.1 | |||
| XM_011531073.2 | XP_011529375.1 | |||
| XM_011531074.2 | XP_011529376.1 | |||
| XM_011531075.2 | XP_011529377.1 | |||
| XM_017029998.1 | XP_016885487.1 | |||
| XM_017029999.1 | XP_016885488.1 | |||
| XM_017030000.1 | XP_016885489.1 | |||
| XM_017030001.1 | XP_016885490.1 | |||
| XM_017030002.1 | XP_016885491.1 | |||
| XM_017030003.1 | XP_016885492.1 | |||
| AB014598.2 | ||||
| AF148860.1 | AAK08131.1 | |||
| AJ296162.2 | CAC35365.2 | |||
| AK307761.1 | ||||
| AK311043.1 | ||||
| AY358990.1 | AAQ89349.1 | |||
| BC011561.1 | AAH11561.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576919 | Chr.X:65837678 - 66804698 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map