Assay Details
Target Gene Details
Entrez Gene ID: | 26580 |
Gene Name: | BSCL2, seipin lipid droplet biogenesis associated |
Gene Aliases: |
GNG3LG, HMN5, PELD, SPG17 |
Location: |
Chr.11:62690262-62709619 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BSCL2 | NR_037948.1 | |||
| NR_037949.1 | ||||
| AK315423.1 | ||||
| BP370169.1 |
Target Gene Details
Entrez Gene ID: | 2785 |
Gene Name: | G protein subunit gamma 3 |
Gene Aliases: |
- |
Location: |
Chr.11:62707255-62709206 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNG3 | NM_012202.4 | 3 | 647 | NP_036334.1 |
| XM_006718500.2 | 4 | 634 | XP_006718563.1 | |
| AF075042.1 | 2 | 310 | ||
| AF087900.1 | 3 | 584 | AAP97199.1 | |
| AF092129.1 | 3 | 584 | AAD40371.1 | |
| BC015563.2 | 3 | 528 | AAH15563.1 | |
| CK004020.1 |
Target Gene Details
Entrez Gene ID: | 100534595 |
Gene Name: | HNRNPUL2-BSCL2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:62690262-62727384 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPUL2-BSCL2 | NR_037946.1 | |||
| AK122942.1 | ||||
| CN346952.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv348 | Chr.11:62706978 - 62752240 on Build GRCh38 | Deletion |
|
| nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map