Assay Details
Target Gene Details
Entrez Gene ID: | 23586 |
Gene Name: | DEXD/H-box helicase 58 |
Gene Aliases: |
RIG-I, RIGI, RLR-1, SGMRT2 |
Location: |
Chr.9:32455302-32526324 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX58 | NM_014314.3 | 18 | 2957 | NP_055129.2 |
| AF038963.1 | 18 | 2956 | AAD19826.1 | |
| AK023661.1 | 11 | 1632 | ||
| AK125989.1 | 19 | 3162 | ||
| AK315040.1 | ||||
| AL137608.1 | 8 | 1218 | CAB70840.1 | |
| BC132786.1 | 18 | 2905 | ||
| BC136610.1 | 18 | 2905 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519447 | Chr.9:32456658 - 32457560 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map