Assay Details
Target Gene Details
Entrez Gene ID: | 79654 |
Gene Name: | HECT domain E3 ubiquitin protein ligase 3 |
Gene Aliases: |
- |
Location: |
Chr.1:45002548-45011355 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HECTD3 | NM_024602.5 | 17 | 2285 | NP_078878.3 |
| XM_011542140.2 | 15 | 1682 | XP_011540442.1 | |
| AK056545.1 | 15 | 1636 | ||
| AK056873.1 | 9 | 1179 | ||
| AK091583.1 | 2 | 1034 | ||
| AK096462.1 | 17 | 2164 | ||
| AK301124.1 | 16 | 1873 | ||
| BC019105.2 | 2 | 244 | ||
| BC126352.1 | 14 | 1562 | ||
| BC130470.1 | 14 | 1562 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950706 | Chr.1:45001329 - 45011128 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map