Assay Details
Target Gene Details
Entrez Gene ID: | 344905 |
Gene Name: | ATPase 13A5 |
Gene Aliases: |
- |
Location: |
Chr.3:193274789-193378824 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP13A5 | NM_198505.2 | NP_940907.2 | ||
| XM_011512770.1 | XP_011511072.1 | |||
| XM_017006304.1 | XP_016861793.1 | |||
| XM_017006305.1 | XP_016861794.1 | |||
| AY823163.1 | AAX24103.1 |
Target Gene Details
Entrez Gene ID: | 100874218 |
Gene Name: | ATP13A5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:193307244-193314521 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP13A5-AS1 | NR_046758.1 | |||
| CB054191.1 | ||||
| CR746107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003505 | Chr.3:193168634 - 193574329 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78188075] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map