Assay Details
Target Gene Details
Entrez Gene ID: | 126074 |
Gene Name: | SWIM-type zinc finger 7 associated protein 1 |
Gene Aliases: |
C19orf39, SWS1AP1, ZSWIM7AP1 |
Location: |
Chr.19:11374707-11376951 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SWSAP1 | NM_175871.3 | NP_787067.2 | ||
| AK092438.1 | BAC03891.1 | |||
| BC068071.1 | AAH68071.1 | |||
| BC119677.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833752 | Chr.19:11208324 - 11448939 on Build GRCh38 | Loss |
|
| nsv519802 | Chr.19:11236817 - 11578343 on Build GRCh38 | Loss |
|
| esv26258 | Chr.19:11374513 - 11375528 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map