Assay Details
Target Gene Details
Entrez Gene ID: | 53 |
Gene Name: | acid phosphatase 2, lysosomal |
Gene Aliases: |
LAP |
Location: |
Chr.11:47239302-47248847 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACP2 | NM_001302489.1 | NP_001289418.1 | ||
| NM_001302490.1 | NP_001289419.1 | |||
| NM_001302491.1 | NP_001289420.1 | |||
| NM_001302492.1 | NP_001289421.1 | |||
| NM_001610.3 | NP_001601.1 | |||
| AK298122.1 | ||||
| AK298439.1 | ||||
| AK299382.1 | ||||
| AK300569.1 | ||||
| AK300911.1 | ||||
| AK300931.1 | ||||
| AK301860.1 | ||||
| AK303894.1 | ||||
| AK308512.1 | ||||
| AK311431.1 | ||||
| BC003160.1 | AAH03160.1 | |||
| BC093010.1 | AAH93010.1 | |||
| DA382854.1 | ||||
| DC337294.1 | ||||
| X12548.1 | CAA31064.1 |
More Information
Set Membership: |
|

Genomic Map