Assay Details
Target Gene Details
Entrez Gene ID: | 26284 |
Gene Name: | Era like 12S mitochondrial rRNA chaperone 1 |
Gene Aliases: |
CEGA, ERA, ERA-W, ERAL1A, H-ERA, HERA-A, HERA-B |
Location: |
Chr.17:28854957-28861067 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERAL1 | NM_001317985.1 | NP_001304914.1 | ||
| NM_001317986.1 | NP_001304915.1 | |||
| NM_005702.3 | NP_005693.1 | |||
| NR_134328.1 | ||||
| AB049388.1 | BAB56112.1 | |||
| AF082657.1 | AAC31603.1 | |||
| AF082658.1 | AAC31604.1 | |||
| AK023342.1 | ||||
| AY007435.1 | AAG12978.1 | |||
| BC019094.2 | AAH19094.1 | |||
| BX537542.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640315 | Chr.17:28854410 - 28866359 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map