Assay Details
Target Gene Details
Entrez Gene ID: | 339896 |
Gene Name: | glutamate decarboxylase like 1 |
Gene Aliases: |
ADC, CSADC, HuADC, HuCSADC |
Location: |
Chr.3:30726009-30894661 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GADL1 | NM_207359.2 | 15 | 2830 | NP_997242.2 |
| XM_017006297.1 | 15 | 3228 | XP_016861786.1 | |
| AK128643.1 | 10 | 2259 | BAC87546.1 |
More Information
Set Membership: |
|

Genomic Map