Assay Details
Target Gene Details
Entrez Gene ID: | 114784 |
Gene Name: | CUB and Sushi multiple domains 2 |
Gene Aliases: |
dJ1007G16.1, dJ1007G16.2, dJ947L8.1 |
Location: |
Chr.1:33513999-34165842 on Build GRCh38 |
Assay Gene Location: | Within Exon 73 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSMD2 | NM_001281956.1 | 71 | 11815 | NP_001268885.1 |
| NM_052896.4 | 70 | 11235 | NP_443128.2 | |
| XM_017000185.1 | 70 | 14631 | XP_016855674.1 | |
| XM_017000186.1 | 71 | 11737 | XP_016855675.1 | |
| XM_017000187.1 | 71 | 11563 | XP_016855676.1 | |
| XM_017000188.1 | 70 | 14369 | XP_016855677.1 | |
| XM_017000190.1 | 64 | 13896 | XP_016855679.1 | |
| XM_017000194.1 | 71 | 11472 | XP_016855683.1 | |
| AB067471.1 | 20 | 3585 | BAB67777.1 | |
| AB212622.1 | 71 | 11815 | BAD97692.1 | |
| AK095627.1 | 13 | 2302 | BAC04593.1 | |
| AY210418.1 | 70 | 11235 | AAO34701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585687 | Chr.1:33404909 - 33620947 on Build GRCh38 | Gain |
|
| esv3585686 | Chr.1:33404224 - 33959487 on Build GRCh38 | Gain |
|

Genomic Map