Assay Details
Target Gene Details
Entrez Gene ID: | 9731 |
Gene Name: | centrosomal protein 104 |
Gene Aliases: |
CFAP256, GlyBP, JBTS25, KIAA0562, ROC22 |
Location: |
Chr.1:3812081-3857387 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP104 | XM_011542473.2 | 1 | 79 | XP_011540775.1 |
| XM_011542475.2 | 1 | 79 | XP_011540777.1 | |
| XM_011542476.2 | 1 | 79 | XP_011540778.1 | |
| XM_011542477.2 | 1 | 79 | XP_011540779.1 | |
| XM_011542478.1 | 1 | 79 | XP_011540780.1 | |
| XM_017002917.1 | 1 | 79 | XP_016858406.1 | |
| DA652315.1 | 1 | 78 |
Target Gene Details
Entrez Gene ID: | 1677 |
Gene Name: | DNA fragmentation factor subunit beta |
Gene Aliases: |
CAD, CPAN, DFF-40, DFF2, DFF40 |
Location: |
Chr.1:3857143-3885429 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DFFB | NM_001282669.1 | 1 | 43 | NP_001269598.1 |
| NM_001320132.1 | 1 | 43 | NP_001307061.1 | |
| NM_001320136.1 | 1 | 43 | NP_001307065.1 | |
| NM_004402.3 | 1 | 43 | NP_004393.1 | |
| NR_104222.1 | 1 | 43 | ||
| NR_135150.1 | 1 | 43 | ||
| NR_135151.1 | 1 | 43 | ||
| NR_135152.1 | 1 | 43 | ||
| XM_011540865.2 | XP_011539167.1 | |||
| XM_017000498.1 | 1 | 167 | XP_016855987.1 | |
| XM_017000499.1 | XP_016855988.1 | |||
| BC048797.1 | AAH48797.1 | |||
| DC368011.1 | 1 | 43 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain |
|
| esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain |
|
| nsv509590 | Chr.1:3826457 - 3902492 on Build GRCh38 | Insertion |
|
| esv3585052 | Chr.1:3817921 - 3928224 on Build GRCh38 | Gain |
|
| nsv545130 | Chr.1:3826568 - 4283888 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map