Assay Details
Target Gene Details
Entrez Gene ID: | 83444 |
Gene Name: | INO80 complex subunit B |
Gene Aliases: |
HMGA1L4, HMGIYL4, IES2, PAP-1BP, PAPA-1, PAPA1, ZNHIT4, hIes2 |
Location: |
Chr.2:74455023-74457960 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80B | NM_031288.3 | NP_112578.2 | ||
| AB054538.1 | BAB21111.1 | |||
| AK295889.1 | ||||
| AK295901.1 | 1 | 41 | ||
| BC050666.2 | ||||
| BC064425.1 | ||||
| CN260703.1 | ||||
| DB451812.1 |
Target Gene Details
Entrez Gene ID: | 100532735 |
Gene Name: | INO80B-WBP1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.2:74455023-74460891 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INO80B-WBP1 | NR_037849.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458185 | Chr.2:74367887 - 74531807 on Build GRCh38 | Loss |
|
| esv2759061 | Chr.2:74252797 - 74535298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map