Assay Details
Target Gene Details
Entrez Gene ID: | 57045 |
Gene Name: | twisted gastrulation BMP signaling modulator 1 |
Gene Aliases: |
TSG |
Location: |
Chr.18:9334767-9402420 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TWSG1 | NM_020648.5 | 3 | 356 | NP_065699.1 |
| AF196834.1 | 3 | 313 | AAG43468.1 | |
| AF294628.1 | 2 | 178 | AAG10076.1 | |
| AJ297391.1 | 3 | 307 | CAC05581.1 | |
| AK027629.1 | 3 | 356 | BAB55246.1 | |
| AK075550.1 | 3 | 356 | BAC11693.1 | |
| AK315751.1 | 3 | 241 | ||
| BC020490.1 | 3 | 270 | AAH20490.1 | |
| DA735800.1 | 3 | 356 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056407 | Chr.18:9144347 - 9553766 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map