Assay Details
Target Gene Details
Entrez Gene ID: | 84708 |
Gene Name: | ligand of numb-protein X 1 |
Gene Aliases: |
LNX, MPDZ, PDZRN2 |
Location: |
Chr.4:53458264-53652524 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LNX1 | NM_001126328.2 | 9 | 2043 | NP_001119800.1 |
| NM_032622.2 | 8 | 1774 | NP_116011.2 | |
| XM_005265785.4 | 9 | 2201 | XP_005265842.1 | |
| XM_017008776.1 | 10 | 2262 | XP_016864265.1 | |
| AF237782.1 | 8 | 1705 | AAK15039.1 | |
| AK056823.1 | 9 | 2016 | BAB71291.1 | |
| AK294611.1 | 9 | 1572 | ||
| AY358326.1 | 8 | 1685 | AAQ88692.1 | |
| BC022983.1 | 9 | 1919 | AAH22983.1 | |
| BC034737.1 | 8 | 1694 | AAH34737.1 |
More Information
Set Membership: |
|

Genomic Map