Assay Details
Target Gene Details
Entrez Gene ID: | 147372 |
Gene Name: | collagen and calcium binding EGF domains 1 |
Gene Aliases: |
HKLLS1 |
Location: |
Chr.18:59430939-59698181 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCBE1 | NM_133459.3 | NP_597716.1 | ||
| XM_017025556.1 | XP_016881045.1 | |||
| XM_017025557.1 | XP_016881046.1 | |||
| XM_017025558.1 | XP_016881047.1 | |||
| AB075863.1 | ||||
| AY358347.1 | AAQ88713.1 | |||
| BC046645.1 | AAH46645.1 | |||
| BX640826.1 | CAE45902.1 |
More Information
Set Membership: |
|

Genomic Map