Assay Details
Target Gene Details
Entrez Gene ID: | 51667 |
Gene Name: | negative regulator of ubiquitin like proteins 1 |
Gene Aliases: |
BS4, NUB1L, NYREN18 |
Location: |
Chr.7:151341699-151378461 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUB1 | NM_001243351.1 | 15 | 2376 | NP_001230280.1 |
| NM_016118.4 | 15 | 2334 | NP_057202.3 | |
| XM_005250011.2 | 15 | 2441 | XP_005250068.1 | |
| XM_017012304.1 | 15 | 2435 | XP_016867793.1 | |
| XM_017012305.1 | 15 | 2393 | XP_016867794.1 | |
| XM_017012306.1 | 15 | 2415 | XP_016867795.1 | |
| XM_017012307.1 | 14 | 2291 | XP_016867796.1 | |
| XM_017012308.1 | 14 | 2288 | XP_016867797.1 | |
| XM_017012309.1 | 14 | 2294 | XP_016867798.1 | |
| XM_017012310.1 | 16 | 2538 | XP_016867799.1 | |
| XM_017012311.1 | 16 | 2442 | XP_016867800.1 | |
| XM_017012312.1 | 16 | 2411 | XP_016867801.1 | |
| XM_017012313.1 | 16 | 2496 | XP_016867802.1 | |
| XM_017012314.1 | 16 | 2372 | XP_016867803.1 | |
| AF300717.1 | 15 | 2323 | AAK21001.1 | |
| AF459743.1 | 15 | 2324 | AAO14547.1 | |
| AY129295.1 | 15 | 2282 | AAN01355.1 | |
| BC046354.2 | 14 | 2250 | AAH46354.1 |
More Information
Set Membership: |
|

Genomic Map