Assay Details
Target Gene Details
Entrez Gene ID: | 2752 |
Gene Name: | glutamate-ammonia ligase |
Gene Aliases: |
GLNS, GS, PIG43, PIG59 |
Location: |
Chr.1:182378093-182392206 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLUL | NM_001033044.3 | 7 | 2184 | NP_001028216.1 |
| NM_001033056.3 | 7 | 1886 | NP_001028228.1 | |
| NM_002065.6 | 8 | 2540 | NP_002056.2 | |
| XM_006711278.1 | 7 | 2247 | XP_006711341.1 | |
| AK301600.1 | ||||
| AL161952.1 | 2 | 4005 | CAB82304.1 | |
| AY947343.1 | 7 | 1744 | ||
| BC011700.2 | 7 | 1752 | AAH11700.1 | |
| BC011852.2 | 7 | 1763 | AAH11852.1 | |
| BC018992.1 | 8 | 2103 | AAH18992.1 | |
| BC051726.1 | 7 | 2949 | AAH51726.1 | |
| BX537384.1 | 7 | 1768 | CAD97626.1 | |
| S70290.1 | 6 | 1643 | AAB30693.1 | |
| X59834.1 | 7 | 1752 | CAA42495.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548358 | Chr.1:182110851 - 183151893 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map