Assay Details
Target Gene Details
Entrez Gene ID: | 6898 |
Gene Name: | tyrosine aminotransferase |
Gene Aliases: |
- |
Location: |
Chr.16:71566851-71577095 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAT | NM_000353.2 | NP_000344.1 | ||
| AK313380.1 | ||||
| BC130534.1 | ||||
| X52520.1 | CAA36750.1 | |||
| X55675.1 | CAA39210.1 |
Target Gene Details
Entrez Gene ID: | 100132529 |
Gene Name: | TAT antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.16:71565016-71572438 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAT-AS1 | NR_103851.1 | |||
| NR_103852.1 | ||||
| AA913333.1 | ||||
| BM853033.1 | ||||
| BU853575.1 | ||||
| BU854532.1 | ||||
| HY017421.1 |
More Information
Set Membership: |
|

Genomic Map