Assay Details
Target Gene Details
Entrez Gene ID: | 55799 |
Gene Name: | calcium voltage-gated channel auxiliary subunit alpha2delta 3 |
Gene Aliases: |
HSA272268 |
Location: |
Chr.3:54122594-55074557 on Build GRCh38 |
Assay Gene Location: | Within Intron 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNA2D3 | NM_018398.2 | NP_060868.2 | ||
| XM_005265318.2 | XP_005265375.1 | |||
| XM_011533946.2 | XP_011532248.1 | |||
| XM_011533947.2 | XP_011532249.1 | |||
| XM_011533954.2 | XP_011532256.1 | |||
| XM_011533955.1 | XP_011532257.1 | |||
| XM_017006850.1 | XP_016862339.1 | |||
| XM_017006851.1 | XP_016862340.1 | |||
| XM_017006853.1 | XP_016862342.1 | |||
| XM_017006854.1 | XP_016862343.1 | |||
| AF516696.1 | AAN06673.1 | |||
| AJ272213.1 | CAB75878.1 | |||
| AJ272268.1 | CAB75962.1 | |||
| BC137502.1 | ||||
| BC137505.1 | ||||
| BI753834.1 |
Target Gene Details
Entrez Gene ID: | 57408 |
Gene Name: | leucine rich repeats and transmembrane domains 1 |
Gene Aliases: |
HT017 |
Location: |
Chr.3:54918333-54967088 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRTM1 | NM_001304389.1 | 3 | 1193 | NP_001291318.1 |
| NM_020678.3 | 3 | 1202 | NP_065729.1 | |
| AF225421.1 | 3 | 1173 | AAG09723.1 | |
| BC040732.1 | 3 | 1193 | AAH40732.1 | |
| BC074741.2 | 3 | 1063 | AAH74741.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517851 | Chr.3:54375499 - 55705332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map