Assay Details
Target Gene Details
Entrez Gene ID: | 57719 |
Gene Name: | anoctamin 8 |
Gene Aliases: |
KIAA1623, TMEM16H |
Location: |
Chr.19:17323223-17334834 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANO8 | NM_020959.2 | NP_066010.1 | ||
| XM_017027048.1 | XP_016882537.1 | |||
| AB046843.2 | ||||
| AI860149.1 |
Target Gene Details
Entrez Gene ID: | 79016 |
Gene Name: | DET1 and DDB1 associated 1 |
Gene Aliases: |
C19orf58, PCIA1 |
Location: |
Chr.19:17309528-17323301 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDA1 | NM_024050.5 | 5 | 4006 | NP_076955.1 |
| AL832922.1 | 1 | 3116 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470129 | Chr.19:17188084 - 17341718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map