Assay Details
Target Gene Details
Entrez Gene ID: | 3712 |
Gene Name: | isovaleryl-CoA dehydrogenase |
Gene Aliases: |
ACAD2 |
Location: |
Chr.15:40405485-40435948 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IVD | NM_001159508.1 | 1 | 191 | NP_001152980.1 |
| NM_002225.3 | 1 | 191 | NP_002216.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510660 | Chr.15:40260705 - 40429684 on Build GRCh38 | Deletion |
|
| esv34145 | Chr.15:40257416 - 40597626 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map