Assay Details
Target Gene Details
Entrez Gene ID: | 55341 |
Gene Name: | large 60S subunit nuclear export GTPase 1 |
Gene Aliases: |
- |
Location: |
Chr.3:194640788-194672477 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LSG1 | NM_018385.2 | 14 | 2900 | NP_060855.2 |
| XM_017006796.1 | 10 | 2228 | XP_016862285.1 | |
| AL136897.1 | 14 | 2605 | CAB66831.1 | |
| AW748476.1 | 1 | 65 | ||
| BC068500.1 | 14 | 2614 | AAH68500.1 | |
| BQ009975.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599208 | Chr.3:194592316 - 194799085 on Build GRCh38 | Loss |
|
| nsv508268 | Chr.3:194618407 - 194681984 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map