Assay Details
Target Gene Details
Entrez Gene ID: | 53343 |
Gene Name: | nudix hydrolase 9 |
Gene Aliases: |
NUDT10 |
Location: |
Chr.4:87420002-87459454 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT9 | NM_001248011.1 | 2 | 592 | NP_001234940.1 |
| NM_024047.4 | 2 | 592 | NP_076952.1 | |
| NM_198038.2 | 2 | 234 | NP_932155.1 | |
| XM_011532031.2 | 2 | 595 | XP_011530333.1 | |
| XM_011532032.2 | 2 | 2771 | XP_011530334.1 | |
| AF265341.1 | 2 | 233 | AAM46066.1 | |
| AF273028.1 | 2 | 556 | AAM46068.1 | |
| AK027295.1 | 2 | 582 | BAB55021.1 | |
| AK074845.1 | 2 | 234 | BAC11239.1 | |
| AK075408.1 | 2 | 512 | BAC11601.1 | |
| AK308028.1 | 2 | 359 | ||
| AY026252.1 | 2 | 587 | AAK07671.1 | |
| AY359123.1 | 2 | 286 | AAQ89480.1 | |
| BC000542.2 | 2 | 352 | AAH00542.1 | |
| BM544951.1 | 2 | 604 | ||
| BP277630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3601276 | Chr.4:87422870 - 87515781 on Build GRCh38 | Loss |
|
| esv3601277 | Chr.4:87430386 - 87443786 on Build GRCh38 | Loss |
|
| nsv1002789 | Chr.4:87318188 - 87685333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map