Assay Details
Target Gene Details
Entrez Gene ID: | 105371932 |
Gene Name: | uncharacterized LOC105371932 |
Gene Aliases: |
- |
Location: |
Chr.17:35240043-35247472 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105371932 | XM_017025492.1 | 2 | 172 | XP_016880981.1 |
Target Gene Details
Entrez Gene ID: | 162394 |
Gene Name: | schlafen family member 5 |
Gene Aliases: |
- |
Location: |
Chr.17:35243026-35273650 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLFN5 | NM_144975.3 | NP_659412.3 | ||
| XM_005257934.3 | XP_005257991.1 | |||
| AK054668.1 | BAB70788.1 | |||
| AK303299.1 | ||||
| AK303632.1 | ||||
| BC021238.1 | AAH21238.1 | |||
| BP281007.1 | ||||
| BX647942.1 | ||||
| CD704489.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422399 | Chr.17:35212221 - 35291582 on Build GRCh38 | Deletion |
|
| esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss |
|
| nsv574741 | Chr.17:35225256 - 35500115 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map