Assay Details
Target Gene Details
Entrez Gene ID: | 221395 |
Gene Name: | adhesion G protein-coupled receptor F5 |
Gene Aliases: |
GPR116, KPG_001 |
Location: |
Chr.6:46852505-46954954 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Target Gene Details
Entrez Gene ID: | 101926962 |
Gene Name: | uncharacterized LOC101926962 |
Gene Aliases: |
- |
Location: |
Chr.6:46903471-46910029 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101926962 | NR_110658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830654 | Chr.6:46755161 - 46938169 on Build GRCh38 | Gain |
|
| nsv428147 | Chr.6:46732445 - 46923866 on Build GRCh38 | Gain |
|
| nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map