Assay Details
Target Gene Details
Entrez Gene ID: | 5653 |
Gene Name: | kallikrein related peptidase 6 |
Gene Aliases: |
Bssp, Klk7, PRSS18, PRSS9, SP59, hK6 |
Location: |
Chr.19:50958631-50969673 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK6 | NM_001012964.2 | NP_001012982.1 | ||
| NM_001012965.2 | NP_001012983.1 | |||
| NM_001319948.1 | NP_001306877.1 | |||
| NM_001319949.1 | NP_001306878.1 | |||
| NM_002774.3 | NP_002765.1 | |||
| AF013988.1 | AAB66483.1 | |||
| AK314897.1 | ||||
| AY279383.1 | ||||
| AY318867.1 | AAP82446.1 | |||
| AY318868.1 | ||||
| AY318869.1 | AAP82448.1 | |||
| AY318870.1 | ||||
| AY457039.1 | AAR23148.1 | |||
| BC015525.2 | AAH15525.1 | |||
| BM763657.1 | ||||
| BP253007.1 | ||||
| BT006852.1 | AAP35498.1 | |||
| D78203.1 | BAA11306.1 | |||
| DQ223012.1 | ABB04464.1 | |||
| U62801.1 | AAB07113.1 |
More Information
Set Membership: |
|

Genomic Map