Assay Details
Target Gene Details
Entrez Gene ID: | 23033 |
Gene Name: | dopey family member 1 |
Gene Aliases: |
DOP1, KIAA1117, dJ202D23.2 |
Location: |
Chr.6:83067650-83171351 on Build GRCh38 |
Assay Gene Location: | Within Exon 39 |
Target Gene Details
Entrez Gene ID: | 5238 |
Gene Name: | phosphoglucomutase 3 |
Gene Aliases: |
AGM1, IMD23, PAGM, PGM 3 |
Location: |
Chr.6:83150728-83193936 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGM3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map