Assay Details
Target Gene Details
Entrez Gene ID: | 27141 |
Gene Name: | cell death-inducing DFFA-like effector b |
Gene Aliases: |
- |
Location: |
Chr.14:24305187-24311377 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CIDEB | NM_001318807.1 | NP_001305736.1 | ||
| NM_014430.2 | NP_055245.2 | |||
| XM_011536659.2 | XP_011534961.1 | |||
| XM_017021221.1 | XP_016876710.1 | |||
| AF190901.1 | AAF23324.1 | |||
| AK122828.1 | 1 | 907 | ||
| BX418750.2 | ||||
| DN997151.1 |
Target Gene Details
Entrez Gene ID: | 56413 |
Gene Name: | leukotriene B4 receptor 2 |
Gene Aliases: |
BLT2, BLTR2, JULF2, KPG_004, LTB4-R 2, LTB4-R2, NOP9 |
Location: |
Chr.14:24310151-24312053 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTB4R2 | NM_001164692.2 | NP_001158164.1 | ||
| NM_019839.4 | NP_062813.2 | |||
| AB032558.1 | BAD83590.1 | |||
| AF277230.1 | 1 | 1590 | AAF81261.1 | |
| AK304169.1 | ||||
| BC119674.2 | 1 | 184 | ||
| BC127643.1 | 1 | 187 | ||
| DC408599.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564058 | Chr.14:24302079 - 24317770 on Build GRCh38 | Loss |
|
| nsv564060 | Chr.14:24307195 - 24340589 on Build GRCh38 | Loss |
|
| esv2669637 | Chr.14:24274158 - 24318169 on Build GRCh38 | Deletion |
|
| esv3633804 | Chr.14:24274134 - 24318147 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map