Assay Details
Target Gene Details
Entrez Gene ID: | 5650 |
Gene Name: | kallikrein related peptidase 7 |
Gene Aliases: |
PRSS6, SCCE, hK7 |
Location: |
Chr.19:50976479-50984064 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK7 | NM_001207053.1 | NP_001193982.1 | ||
| NM_001243126.1 | NP_001230055.1 | |||
| NM_005046.3 | NP_005037.1 | |||
| AF411214.1 | AAN03662.1 | |||
| AF411215.1 | AAN03663.1 | |||
| AK289660.1 | ||||
| AK295313.1 | ||||
| BC032005.1 | AAH32005.1 | |||
| DA243628.1 | ||||
| DA307925.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064207 | Chr.19:50971269 - 50994022 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map