Assay Details
Target Gene Details
Entrez Gene ID: | 57650 |
Gene Name: | KIAA1524 |
Gene Aliases: |
CIP2A, p90 |
Location: |
Chr.3:108545752-108589644 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA1524 | NM_020890.2 | 21 | 2897 | NP_065941.2 |
| XM_006713716.3 | 21 | 2681 | XP_006713779.1 | |
| XM_006713717.2 | 22 | 2793 | XP_006713780.1 | |
| XM_011513056.1 | 21 | 2641 | XP_011511358.1 | |
| XM_011513057.2 | 22 | 2762 | XP_011511359.1 | |
| AB040957.1 | 21 | 2624 | BAA96048.1 | |
| AF334474.1 | 21 | 2696 | AAL11632.1 | |
| AK022912.1 | 12 | 1508 | BAB14305.1 | |
| AK308315.1 | 21 | 2644 | ||
| AL713689.1 | 21 | 2692 | CAD28489.1 | |
| BC017659.1 | 7 | 770 | AAH17659.1 | |
| BC130564.1 | 21 | 2664 | ||
| BC136371.1 | 21 | 2664 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829658 | Chr.3:108484408 - 108666131 on Build GRCh38 | Gain |
|
| nsv1011158 | Chr.3:107746970 - 108629120 on Build GRCh38 | Gain |
|
| nsv1006774 | Chr.3:108522528 - 108559230 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map