Assay Details
Target Gene Details
Entrez Gene ID: | 101927969 |
Gene Name: | uncharacterized LOC101927969 |
Gene Aliases: |
- |
Location: |
Chr.5:168706927-168720884 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927969 | NR_130737.1 | |||
| AK126009.1 |
Target Gene Details
Entrez Gene ID: | 6586 |
Gene Name: | slit guidance ligand 3 |
Gene Aliases: |
MEGF5, SLIL2, SLIT1, Slit-3, slit2 |
Location: |
Chr.5:168661733-169301129 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 24 - Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLIT3 | NM_001271946.1 | NP_001258875.1 | ||
| NM_003062.3 | NP_003053.1 | |||
| XM_017009779.1 | XP_016865268.1 | |||
| AB011538.2 | BAA32466.2 | |||
| AB017169.1 | BAA35186.1 | |||
| AF075240.1 | AAD19336.1 | |||
| AY358884.1 | AAQ89243.1 | |||
| BC146759.1 |
More Information
Set Membership: |
|

Genomic Map