Assay Details
Target Gene Details
Entrez Gene ID: | 27245 |
Gene Name: | AT-hook DNA binding motif containing 1 |
Gene Aliases: |
MRD25 |
Location: |
Chr.1:27534245-27604178 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHDC1 | NM_001029882.3 | 6 | 5363 | NP_001025053.1 |
| XM_005245848.3 | 7 | 5598 | XP_005245905.1 | |
| XM_005245849.3 | 7 | 5703 | XP_005245906.1 | |
| XM_005245850.3 | 8 | 5723 | XP_005245907.1 | |
| XM_005245851.3 | 7 | 5595 | XP_005245908.1 | |
| XM_005245852.3 | 6 | 5372 | XP_005245909.1 | |
| XM_011541255.1 | 7 | 5277 | XP_011539557.1 | |
| XM_011541256.2 | 7 | 5717 | XP_011539558.1 | |
| XM_011541257.2 | 6 | 6472 | XP_011539559.1 | |
| AK125431.1 | 1 | 2081 | BAC86163.1 | |
| BC002677.2 | 1 | 1159 | AAH02677.2 | |
| BC014394.1 | 1 | 1159 | ||
| U79259.1 | 1 | 1125 | AAB50205.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv14n27 | Chr.1:27445496 - 27618848 on Build GRCh38 | Loss |
|
| nsv520790 | Chr.1:27532525 - 27549971 on Build GRCh38 | Loss |
|
| esv6834 | Chr.1:27547531 - 27547681 on Build GRCh38 | Gain |
|
| dgv219n54 | Chr.1:27459823 - 27618848 on Build GRCh38 | Loss |
|
| dgv220n54 | Chr.1:27522789 - 27631734 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map