Assay Details
Target Gene Details
Entrez Gene ID: | 51691 |
Gene Name: | LSM8 homolog, U6 small nuclear RNA associated |
Gene Aliases: |
NAA38 |
Location: |
Chr.7:118184032-118204039 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LSM8 | NM_016200.4 | 4 | 928 | NP_057284.1 |
| AL600056.1 | 1 | 211 | ||
| BC022440.1 | 4 | 798 | AAH22440.1 | |
| BM972935.1 | 1 | 396 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv608259 | Chr.7:117837974 - 119521689 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map